Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((1H-Imidazol-1-yl)methyl)benzoic acid 97%

Supplier: AMBEED, INC
Warning

A258267-1G A258267-250MG
77238-432EA 261 USD InStock
77238-432 77238-434
2-((1H-Imidazol-1-yl)methyl)benzoic acid 97%
2-((1H-Imidazol-1-yl)methyl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₂O₂
MDL Number: MFCD07186452
CAS Number: 883543-82-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty