Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-1H-imidazole-4,5-dicarbonitrile 97%

Supplier: AMBEED, INC
Warning

A268489-10G A268489-1G A268489-25G
77250-282EA 20 USD InStock
77250-282 77250-284 77250-286
1-Ethyl-1H-imidazole-4,5-dicarbonitrile 97%
1-Ethyl-1H-imidazole-4,5-dicarbonitrile
Precaución: for Research Use Only


Formula: C₇H₆N₄
MDL Number: MFCD06762788
CAS Number: 133123-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty