Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Isoquinolinol 95%

Supplier: AMBEED, INC
Warning

A270646-1G A270646-5G
77262-446EA 12.36 USD InStock
77262-446 77262-448 77262-450
7-Isoquinolinol 95%
7-Isoquinolinol
Precaución: for Research Use Only


Formula: C₉H₇NO
MDL Number: MFCD00456131
CAS Number: 7651-83-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty