Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-chlorobenzamide 98%

Supplier: AMBEED, INC
Warning

A287942-1G A287942-5G
77277-128EA 15 USD InStock
77277-128 77277-130
3-Bromo-5-chlorobenzamide 98%
3-Bromo-5-chlorobenzamide
Precaución: for Research Use Only


Formula: C₇H₅BrClNO
MDL Number: MFCD07780655
CAS Number: 933671-77-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty