Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tafenoquine 97%

Supplier: AMBEED, INC
Warning

A299706-1MG A299706-5MG
77262-974EA 25 USD InStock
77262-974 77262-976
Tafenoquine 97%
Tafenoquine
Precaución: for Research Use Only


Formula: C₂₄H₂₈F₃N₃O₃
MDL Number: MFCD00598712
CAS Number: 106635-80-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty