Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-((3-((4-(Morpholinomethyl)-1H-pyrrol-2-yl)methylene)-2-oxoindolin-5-yl)methyl)thiazolidine-2,4-dione 98%

Supplier: AMBEED, INC
Warning

A300808-10MG A300808-1MG A300808-2MG A300808-50MG A300808-5MG
77262-990EA 70 USD InStock
77262-990 77262-992 77262-994 77262-996 77262-998
3-((3-((4-(Morpholinomethyl)-1H-pyrrol-2-yl)methylene)-2-oxoindolin-5-yl)methyl)thiazolidine-2,4-dione 98%
3-((3-((4-(Morpholinomethyl)-1H-pyrrol-2-yl)methylene)-2-oxoindolin-5-yl)methyl)thiazolidine-2,4-dione
Precaución: for Research Use Only


Formula: C₂₂H₂₂N₄O₄S
MDL Number: MFCD28963943
CAS Number: 1265965-22-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty