Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1-Naphthyl)-D-alanine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A301197-1G A301197-250MG A301197-25G A301197-5G
77277-318EA 17 USD InStock
77277-318 77277-320 77277-322 77277-324
3-(1-Naphthyl)-D-alanine hydrochloride 95%
3-(1-Naphthyl)-D-alanine hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₁₃NO₂
MW: 251.71 g/mol
MDL Number: MFCD03095597
CAS Number: 122745-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty