Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitro-m-anisic acid 98%

Supplier: AMBEED, INC
Warning

A307315-100G A307315-10G A307315-1G A307315-25G A307315-5G
77239-344EA 294 USD InStock
77239-344 77239-346 77239-348 77239-350 77239-352
2-Nitro-m-anisic acid 98%
2-Nitro-m-anisic acid
Precaución: for Research Use Only


Formula: C₈H₇NO₅
MDL Number: MFCD00007143
CAS Number: 4920-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty