Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dihexyl carbonate 97%

Supplier: AMBEED, INC
Warning

A307374-100MG A307374-1G A307374-250MG A307374-5G
77263-142EA 57 USD InStock
77263-142 77263-144 77263-146 77263-148
Dihexyl carbonate 97%
Dihexyl carbonate
Precaución: for Research Use Only


Formula: C₁₃H₂₆O₃
MDL Number: MFCD00068965
CAS Number: 7523-15-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty