(Bromoethynyl)triisopropylsilane 98%

Supplier: AMBEED, INC
Warning

A320630-100G A320630-1G A320630-250MG A320630-25G A320630-5G
77251-128EA 243 USD
77251-128 77251-130 77251-132 77251-134 77251-136 77251-138
(Bromoethynyl)triisopropylsilane 98%
(Bromoethynyl)triisopropylsilane

Precaución: for Research Use Only

Formula: C₁₁H₂₁BrSi
MW: 261.28 g/mol
MDL Number: MFCD11036302
CAS Number: 111409-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR