2-Chloro-5-iodobenzotrifluoride 97%
Supplier: AMBEED, INC
|
Warning
|
A322821-10G
A322821-25G
A322821-5G
77263-448EA
11
USD
77263-448
77263-450
77263-452
2-Chloro-5-iodobenzotrifluoride 97%
2-Chloro-5-iodobenzotrifluoride
Precaución: for Research Use Only
Formula:
C₇H₃ClF₃I MW: 306.4514 g/mol |
MDL Number:
MFCD09037801 CAS Number: 260355-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...