4'-Bromo-2,2':6',2''-terpyridine 97%
Supplier: AMBEED, INC
|
Warning
|
A327760-1MG
A327760-250MG
77290-378EA
5
USD
77290-378
77290-380
4'-Bromo-2,2':6',2''-terpyridine 97%
4'-Bromo-2,2':6',2''-terpyridine
Precaución: For research use only.
Formula:
C₁₅H₁₀BrN₃ MW: 312.17 g/mol |
MDL Number:
MFCD06796965 CAS Number: 149817-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...