Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate 98%

Supplier: AMBEED, INC
Warning

A364895-1G A364895-25G A364895-5G
77264-046EA 14 USD InStock
77264-046 77264-048 77264-050
(S)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate 98%
(S)-N,N-Dimethyl-1-ferrocenylethylamine (L)-tartrate
Precaución: for Research Use Only


Formula: C₁₈H₁₆FeNO₆
MDL Number: MFCD09953452
CAS Number: 111614-65-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty