Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A393571-100G A393571-10G A393571-25G A393571-500G A393571-5G
77252-052EA 24 USD InStock
77252-052 77252-054 77252-056 77252-058 77252-060 77252-062
2,5-Dimethylbenzoic acid 98%
2,5-Dimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MDL Number: MFCD00002482
CAS Number: 610-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty