Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole 98%

Supplier: AMBEED, INC
Warning

A396815-100G A396815-1G A396815-250MG A396815-25G A396815-5G
77240-578EA 235 USD InStock
77240-578 77240-580 77240-582 77240-584 77240-586
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole 98%
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₈H₉N₃O
MDL Number: MFCD00051954
CAS Number: 36140-83-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty