Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-5-(4-(3,5-Dimethylisoxazol-4-yl)-2-(pyrrolidin-3-yloxy)phenyl)-2-ureidothiophene-3-carboxamide 98%

Supplier: AMBEED, INC
Warning

A398439-10MG A398439-1MG A398439-25MG A398439-2MG A398439-50MG A398439-5MG
77240-588EA 189 USD InStock
77240-588 77240-590 77240-592 77240-594 77240-596 77240-598
(S)-5-(4-(3,5-Dimethylisoxazol-4-yl)-2-(pyrrolidin-3-yloxy)phenyl)-2-ureidothiophene-3-carboxamide 98%
(S)-5-(4-(3,5-Dimethylisoxazol-4-yl)-2-(pyrrolidin-3-yloxy)phenyl)-2-ureidothiophene-3-carboxamide
Precaución: for Research Use Only


Formula: C₂₁H₂₃N₅O₄S
MDL Number: MFCD29472277
CAS Number: 1609281-86-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty