Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hexylthiophene 98%

Supplier: AMBEED, INC
Danger

A415178-10G A415178-1G A415178-25G A415178-5G
77278-904EA 31 USD InStock
77278-904 77278-906 77278-908 77278-910
3-Hexylthiophene 98%
3-Hexylthiophene
Precaución: for Research Use Only


Formula: C₁₀H₁₆S
MW: 168.3 g/mol
MDL Number: MFCD00143181
CAS Number: 1693-86-3
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty