Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Bis(benzyloxy)-5-bromopyrimidine 96%

Supplier: AMBEED, INC
Warning

A423783-10G A423783-1G A423783-25G A423783-5G
77240-834EA 100 USD InStock
77240-834 77240-836 77240-838 77240-840
2,4-Bis(benzyloxy)-5-bromopyrimidine 96%
2,4-Bis(benzyloxy)-5-bromopyrimidine
Precaución: for Research Use Only


Formula: C₁₈H₁₅BrN₂O₂
MDL Number: MFCD00267932
CAS Number: 41244-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty