tert-Butyl 2-bromo-4-((tert-butoxycarbonyl)amino)nicotinate 97%
Supplier: AMBEED, INC
|
Warning
|
A426430-100MG
A426430-1G
A426430-250MG
77264-796EA
82
USD
77264-796
77264-798
77264-800
tert-Butyl 2-bromo-4-((tert-butoxycarbonyl)amino)nicotinate 97%
tert-Butyl 2-bromo-4-((tert-butoxycarbonyl)amino)nicotinate
Precaución: for Research Use Only
Formula:
C₁₅H₂₁BrN₂O₄ MW: 373.24 g/mol |
MDL Number:
MFCD29905556 CAS Number: 1044148-94-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...