Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-5-pyridineboronic acid 96%

Supplier: AMBEED, INC
Warning

A430583-1G A430583-250MG A430583-25G A430583-5G
77264-856EA 13 USD InStock
77264-856 77264-858 77264-860 77264-862
2-Benzyloxy-5-pyridineboronic acid 96%
2-Benzyloxy-5-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₂BNO₃
MDL Number: MFCD06798262
CAS Number: 929250-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty