Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-amino-2-fluoro-4-methoxybenzoate 97%

Supplier: AMBEED, INC
Danger

A444195-100MG A444195-1G A444195-250MG
77240-988EA 11 USD InStock
77240-988 77240-990 77240-992
Methyl 5-amino-2-fluoro-4-methoxybenzoate 97%
Methyl 5-amino-2-fluoro-4-methoxybenzoate
Precaución: for Research Use Only


Formula: C₉H₁₀FNO₃
MDL Number: MFCD23378835
CAS Number: 1785259-87-1
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty