Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1aR,4aR,6S,7S,7aR,8R,10R,11aS,E)-4a,7,8,10-Tetrahydroxy-1,1,3,6-tetramethyl-9-methylene-1,1a,4a,5,6,7,7a,8,9,10,11,11a-dodecahydro-4H-cyclopenta[a]cyclopropa[f][11]annulen-4-one 97%

Supplier: AMBEED, INC
Warning

A450930-100MG A450930-10MG A450930-1MG A450930-25MG A450930-50MG A450930-5MG
77252-782EA 88 USD InStock
77252-782 77252-784 77252-786 77252-788 77252-790 77252-792
(1aR,4aR,6S,7S,7aR,8R,10R,11aS,E)-4a,7,8,10-Tetrahydroxy-1,1,3,6-tetramethyl-9-methylene-1,1a,4a,5,6,7,7a,8,9,10,11,11a-dodecahydro-4H-cyclopenta[a]cyclopropa[f][11]annulen-4-one 97%
(1aR,4aR,6S,7S,7aR,8R,10R,11aS,E)-4a,7,8,10-Tetrahydroxy-1,1,3,6-tetramethyl-9-methylene-1,1a,4a,5,6,7,7a,8,9,10,11,11a-dodecahydro-4H-cyclopenta[a]cyclopropa[f][11]annulen-4-one
Precaución: for Research Use Only


Formula: C₂₀H₃₀O₅
MDL Number: MFCD30537534
CAS Number: 34208-98-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty