Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MK-8617 97%

Supplier: AMBEED, INC
Warning

A460721-10MG A460721-1MG A460721-5MG A460721-50MG
77265-212EA 36 USD InStock
77265-212 77265-214 77265-216 77748-933
MK-8617 97%
MK-8617
Precaución: for Research Use Only


Formula: C₂₄H₂₁N₅O₄
MDL Number: MFCD22572348
CAS Number: 1187990-87-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty