5-Nitro-2-(morpholin-4-yl)benzoic acid 95%

Supplier: AMBEED, INC
Warning

A461235-100MG A461235-1G A461235-250MG A461235-5G
77279-270EA 5 USD
77279-270 77279-272 77279-274 77748-935
5-Nitro-2-(morpholin-4-yl)benzoic acid 95%
5-Nitro-2-(morpholin-4-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₅
MW: 252.23 g/mol
MDL Number: MFCD00207115
CAS Number: 4036-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR