Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dithiobis(pyridine-n-oxide) 98%

Supplier: AMBEED, INC
Warning

A465705-100G A465705-25G A465705-500G A465705-5G
77252-990EA 240 USD InStock
77252-990 77252-992 77252-994 77252-996
2,2-Dithiobis(pyridine-n-oxide) 98%
2,2-Dithiobis(pyridine-n-oxide)
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O₂S₂
MDL Number: MFCD00006197
CAS Number: 3696-28-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty