Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Bis((2R,5R)-2,5-dimethylphospholano)ferrocene 97%

Supplier: AMBEED, INC
Warning

A477082-100MG A477082-1G A477082-250MG A477082-25G A477082-5G
77253-090EA 22 USD InStock
77253-090 77253-092 77253-094 77253-096 77253-098
1,1-Bis((2R,5R)-2,5-dimethylphospholano)ferrocene 97%
1,1-Bis((2R,5R)-2,5-dimethylphospholano)ferrocene
Precaución: for Research Use Only


Formula: C₂₂H₃₂FeP₂
MDL Number: MFCD08705241
CAS Number: 540475-45-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty