Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,6,6-Tetramethyl-4-piperidone monohydrate 98+%

Supplier: AMBEED, INC
Warning

A477803-100G A477803-500G A477803-25G
77253-100EA 36 USD InStock
77253-100 77253-102 77746-984
2,2,6,6-Tetramethyl-4-piperidone monohydrate 98+%
2,2,6,6-Tetramethyl-4-piperidone monohydrate
Precaución: for Research Use Only


Formula: C₉H₁₇NO·1H₂O
MDL Number: MFCD00149393
CAS Number: 10581-38-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty