Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5aS,10bR)-2-Mesityl-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A480737-100MG A480737-250MG A480737-50MG
77253-128EA 28 USD InStock
77253-128 77253-130 77253-132
(5aS,10bR)-2-Mesityl-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate ≥98%, ee 99%
(5aS,10bR)-2-Mesityl-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₂₁H₂₂BF₄N₃O
MDL Number: MFCD30489133
CAS Number: 1061311-82-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty