Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)-5-nitropyridine 97%

Supplier: AMBEED, INC
Warning

A508991-100MG A508991-1G A508991-250MG
77253-390EA 18 USD InStock
77253-390 77253-392 77253-394
2-(Trifluoromethyl)-5-nitropyridine 97%
2-(Trifluoromethyl)-5-nitropyridine
Precaución: for Research Use Only


Formula: C₆H₃F₃N₂O₂
MDL Number: MFCD07777204
CAS Number: 116470-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty