Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-5-sulfamoylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A523488-100G A523488-25G A523488-5G A523488-500G
77253-522EA 81 USD InStock
77253-522 77253-524 77253-526 77747-069
2-Methoxy-5-sulfamoylbenzoic acid 98%
2-Methoxy-5-sulphamoylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₉NO₅S
MDL Number: MFCD00129997
CAS Number: 22117-85-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty