Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-(2-methoxyethoxy)ethane 90% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)

A551995-100G A551995-1G A551995-25G A551995-5G A551995-500G
77241-974EA 176 USD InStock
77241-974 77241-976 77241-978 77241-980 77747-133
1-Bromo-2-(2-methoxyethoxy)ethane 90% stabilised
1-Bromo-2-(2-methoxyethoxy)ethane
Precaución: for Research Use Only


Formula: BrH₂CCH₂OCH₂CH₂OCH₃
MW: 183.05 g/mol
Boiling Pt: 172
Melting Pt: -23
MDL Number: MFCD00000238
CAS Number: 54149-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty