Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(((4-Methoxy-3-methylpyridin-2-yl)methyl)sulfinyl)-6-(1H-pyrrol-1-yl)-1H-benzo[d]imidazole 97%

Supplier: AMBEED, INC
Warning

A570627-10MG A570627-2MG A570627-50MG A570627-5MG A570627-5G A570627-1G
77242-244EA 14 USD InStock
77242-244 77242-246 77242-248 77242-250 77747-177 77747-176
2-(((4-Methoxy-3-methylpyridin-2-yl)methyl)sulfinyl)-6-(1H-pyrrol-1-yl)-1H-benzo[d]imidazole 97%
2-(((4-Methoxy-3-methylpyridin-2-yl)methyl)sulfinyl)-6-(1H-pyrrol-1-yl)-1H-benzo[d]imidazole
Precaución: for Research Use Only


Formula: C₁₉H₁₈N₄O₂S
MDL Number: MFCD00942538
CAS Number: 172152-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty