Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,7-Dioctylbenzo[b]benzo[4,5]thieno[2,3-d]thiophene 98%

Supplier: AMBEED, INC
Warning

A573973-100MG A573973-250MG
77254-088EA 49 USD InStock
77254-088 77254-090
2,7-Dioctylbenzo[b]benzo[4,5]thieno[2,3-d]thiophene 98%
2,7-Dioctylbenzo[b]benzo[4,5]thieno[2,3-d]thiophene
Precaución: for Research Use Only


Formula: C₃₀H₄₀S₂
MDL Number: MFCD28016330
CAS Number: 583050-70-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty