Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-3-Bromocyclohexene 98% stabilised

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with propylene oxide 300-1000 ppm

A620277-1G A620277-5G
77280-348EA 14 USD InStock
77280-348 77280-350
(±)-3-Bromocyclohexene 98% stabilised
(±)-3-Bromocyclohexene
Precaución: for Research Use Only


Formula: C₆H₉Br
MDL Number: MFCD00013775
CAS Number: 1521-51-3
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty