Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Methylpiperidin-1-yl)ethylamine 98%

Supplier: AMBEED, INC
Warning

A620315-100MG A620315-1G A620315-250MG A620315-50MG
77242-778EA 36 USD InStock
77242-778 77242-780 77242-782 77242-784
2-(4-Methylpiperidin-1-yl)ethylamine 98%
2-(4-Methylpiperidin-1-yl)ethylamine
Precaución: for Research Use Only


Formula: C₈H₁₈N₂
MDL Number: MFCD00129007
CAS Number: 14156-95-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty