Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromophenethoxy)tetrahydro-2H-pyran 97%

Supplier: AMBEED, INC
Warning

A625429-100MG A625429-250MG A625429-5G A625429-1G
77254-460EA 47 USD InStock
77254-460 77254-462 77254-464 77747-289
2-(4-Bromophenethoxy)tetrahydro-2H-pyran 97%
2-(4-Bromophenethoxy)tetrahydro-2H-pyran
Precaución: for Research Use Only


Formula: C₁₃H₁₇BrO₂
MDL Number: MFCD26227285
CAS Number: 79849-46-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty