Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-((N-(4-(tert-Butyl)benzyl)pyridine-3-sulfonamido)methyl)phenoxy)acetic acid 98%

Supplier: AMBEED, INC
Warning

A684831-10MG A684831-1MG A684831-25MG A684831-5MG
77243-332EA 74 USD InStock
77243-332 77243-334 77243-336 77243-338
2-(3-((N-(4-(tert-Butyl)benzyl)pyridine-3-sulfonamido)methyl)phenoxy)acetic acid 98%
2-(3-((N-(4-(tert-Butyl)benzyl)pyridine-3-sulfonamido)methyl)phenoxy)acetic acid
Precaución: for Research Use Only


Formula: C₂₅H₂₈N₂O₅S
MW: 468.56 g/mol
MDL Number: MFCD09833940
CAS Number: 223488-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty