Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1,1-Dioxido-1,2-thiazinan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide 98%

Supplier: AMBEED, INC
Warning

A714403-10MG A714403-1MG A714403-25MG A714403-50MG A714403-5MG
77243-604EA 60 USD InStock
77243-604 77243-606 77243-608 77243-610 77243-612
2-(1,1-Dioxido-1,2-thiazinan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide 98%
2-(1,1-Dioxido-1,2-thiazinan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide
Precaución: for Research Use Only


Formula: C₁₇H₁₉FN₄O₅S
MDL Number: MFCD18074518
CAS Number: 729607-74-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty