2-(Methylamino)phenol 98%

Supplier: AMBEED, INC
Warning

A802424-250MG A802424-5G A802424-1G A802424-25G
77281-398EA 14 USD
77281-398 77281-400 77618-240 77749-589
2-(Methylamino)phenol 98%
2-(Methylamino)phenol

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.152 g/mol
MDL Number: MFCD00462180
CAS Number: 611-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR