Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-fluoro-5-(trifluoromethyl)anisole 98%

Supplier: AMBEED, INC
Warning

A833522-100MG A833522-1G A833522-250MG
77281-606EA 23 USD InStock
77281-606 77281-608 77281-610
3-Chloro-2-fluoro-5-(trifluoromethyl)anisole 98%
3-Chloro-2-fluoro-5-(trifluoromethyl)anisole
Precaución: for Research Use Only


Formula: C₈H₅ClF₄O
MDL Number: MFCD01631517
CAS Number: 261763-01-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty