Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl morpholin-2-ylmethylcarbamate 98%

Supplier: AMBEED, INC
Warning

A839662-1G A839662-250MG A839662-5G
77281-654EA 43 USD InStock
77281-654 77281-656 77281-658
tert-Butyl morpholin-2-ylmethylcarbamate 98%
tert-Butyl morpholin-2-ylmethylcarbamate
Precaución: for Research Use Only


Formula: C₁₀H₂₀N₂O₃
MDL Number: MFCD06808588
CAS Number: 173341-02-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty