Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichlorobis[3-(di-i-propylphosphino)propylamine]ruthenium(II) 98%

Supplier: AMBEED, INC
Warning

A855955-100MG A855955-1G A855955-250MG
77256-332EA 32 USD InStock
77256-332 77256-334 77256-336
Dichlorobis[3-(di-i-propylphosphino)propylamine]ruthenium(II) 98%
Dichlorobis[3-(di-i-propylphosphino)propylamine]ruthenium(II)
Precaución: for Research Use Only


Formula: C₁₈H₄₄Cl₂N₂P₂Ru
MDL Number: MFCD17018813
CAS Number: 1196147-57-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty