Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Pyridyl)morpholine 98%

Supplier: AMBEED, INC
Warning

A863480-10G A863480-1G A863480-25G A863480-5G
77294-052EA 68 USD InStock
77294-052 77294-054 77294-056 77294-058
4-(4-Pyridyl)morpholine 98%
4-(4-Pyridyl)morpholine
Precaución: for Research Use Only


Formula: C₉H₁₂N₂O
MDL Number: MFCD00704054
CAS Number: 2767-91-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty