Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Methoxybenzyl)piperidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A887578-1G A887578-25G A887578-5G
77294-148EA 13 USD InStock
77294-148 77294-150 77294-152
4-(4-Methoxybenzyl)piperidine hydrochloride 95%
4-(4-Methoxybenzyl)piperidine hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₁₉NO·ClH
MDL Number: MFCD03840137
CAS Number: 37581-27-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty