Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3-methoxyphenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A901098-100MG A901098-1G A901098-250MG A901098-5G
77294-170EA 16 USD InStock
77294-170 77294-172 77294-174 77294-176
4-Amino-3-methoxyphenylboronic acid pinacol ester 98%
4-Amino-3-methoxyphenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₂₀BNO₃
MDL Number: MFCD06795657
CAS Number: 461699-81-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty