Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(Benzyloxy)-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: AMBEED, INC
Warning

A945031-1G A945031-250MG A945031-5G
77244-880EA 89 USD InStock
77244-880 77244-882 77244-884
2-(4-(Benzyloxy)-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(4-(Benzyloxy)-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₂₀H₂₅BO₃
MDL Number: MFCD18383421
CAS Number: 1257078-80-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty