Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BBT-594 98+%

Supplier: AMBEED, INC
Warning

A984305-10MG A984305-1MG A984305-25MG A984305-50MG
77269-146EA 76 USD InStock
77269-146 77269-148 77269-150 77269-152
BBT-594 98+%
BBT-594
Precaución: for Research Use Only


Formula: C₂₈H₃₀F₃N₇O₃
MDL Number: MFCD30533519
CAS Number: 882405-89-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty