Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3aR,3'aR,8aS,8'aS)-2,2'-(1-Methylethylidene)bis[3a,8a-dihydro-8H-Indeno[1,2-d]oxazole ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A987510-100MG A987510-1G A987510-250MG
77256-982EA 25 USD InStock
77256-982 77256-984 77256-986
(3aR,3'aR,8aS,8'aS)-2,2'-(1-Methylethylidene)bis[3a,8a-dihydro-8H-Indeno[1,2-d]oxazole ≥98%, ee 99%
(3aR,3'aR,8aS,8'aS)-2,2'-(1-Methylethylidene)bis[3a,8a-dihydro-8H-Indeno[1,2-d]oxazole
Precaución: for Research Use Only


Formula: C₂₃H₂₂N₂O₂
MDL Number: MFCD31707550
CAS Number: 189623-45-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty