Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-MHC Class II beta Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A93079-100 A93079-50
77204-750EA 335 USD InStock
77204-750 77207-060
Anti-MHC Class II beta Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MHC Class II beta for WB with samples derived from Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500-1:2, 000

Type: Primary
Antigen: MHC Class II beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty