Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL-22 Recombinant Recombinant Antibody [clone: DMC471]

Supplier: ANTIBODIES.COM LLC
A318719-100 A318719-500
77187-514EA 703.3 USD InStock
77187-514 77187-516
Anti-IL-22 Recombinant Recombinant Antibody [clone: DMC471]
Antibodies
Recombinant chimeric monoclonal [DMC471] antibody to IL-22 for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: IL-22
Clonality: Recombinant
Clone: DMC471
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty